siRNA Details
Gene Symbol: ATXN1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137675.3 | NP_001131147.1 | 2 | 145 | ||
GenBank mRNA | AB621810.1 | - | 2 | 160 | ||
DB097328.1 | - | 2 | 147 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
