siRNA Details
Gene Symbol: ZNF284 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037813.2 | NP_001032902.1 | 5 | 551 | ||
XM_011526907.2 | XP_011525209.1 | 5 | 592 | |||
XM_011526908.2 | XP_011525210.1 | 6 | 652 | |||
GenBank mRNA | AK308295.1 | - | 11 | 2260 | ||
AY166789.1 | - | 3 | 253 | |||
AY458845.1 | - | 5 | 551 | |||
CR936662.1 | - | 5 | 494 |
