siRNA Details
Gene Symbol: NBPF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047980.2 | NP_001041445.1 | 4 | 947 | ||
XM_017001186.1 | XP_016856675.1 | 4 | 623 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: NBPF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047980.2 | NP_001041445.1 | 4 | 947 | ||
XM_017001186.1 | XP_016856675.1 | 4 | 623 |