siRNA Details
Gene Symbol: PRSS48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183375.2 | NP_899231.2 | 3 | 259 | ||
XM_011531921.2 | XP_011530223.1 | 2 | 705 | |||
GenBank mRNA | BC121109.1 | - | 3 | 263 | 40120369 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
