siRNA Details
Gene Symbol: HSD17B13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136230.2 | NP_001129702.1 | 6 | 798 | ||
NM_178135.4 | NP_835236.2 | 7 | 906 | |||
GenBank mRNA | AB073347.1 | - | 7 | 875 | ||
AK292784.1 | - | 7 | 880 | |||
AY186249.1 | - | 7 | 886 | |||
AY186250.1 | - | 6 | 778 | |||
AY268355.1 | - | 7 | 856 | |||
AY358575.1 | - | 7 | 906 | |||
BC112303.1 | - | 7 | 865 | 8327771 | ||
BC112305.1 | - | 7 | 865 | 8327773 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
