siRNA Details
Gene Symbol: HSD17B13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136230.2 | NP_001129702.1 | 1 | 226 | ||
NM_178135.4 | NP_835236.2 | 1 | 226 | |||
GenBank mRNA | AB073347.1 | - | 1 | 195 | ||
AK292784.1 | - | 1 | 200 | |||
AY186249.1 | - | 1 | 206 | |||
AY186250.1 | - | 1 | 206 | |||
AY268355.1 | - | 1 | 176 | |||
AY358575.1 | - | 1 | 226 | |||
BC112303.1 | - | 1 | 185 | 8327771 | ||
BC112305.1 | - | 1 | 185 | 8327773 | ||
DR004209.1 | - | 1 | 231 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
