siRNA Details
Gene Symbol: MTX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010891.4 | NP_001010891.4 | 2 | 135 | ||
XM_005248495.3 | XP_005248552.1 | 2 | 137 | |||
GenBank mRNA | AK091375.1 | - | 1 | 607 | ||
AK296864.1 | - | 2 | 130 | |||
BC150295.1 | - | 2 | 125 | |||
BX647596.1 | - | 2 | 132 | |||
DB456588.1 | - | 2 | 135 |
