siRNA Details
Gene Symbol: SKA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182620.3 | NP_872426.1 | 3 | 412 | ||
XM_011524745.2 | XP_011523047.1 | 3 | 226 | |||
GenBank mRNA | AK056473.1 | - | 3 | 164 | ||
BC017873.1 | - | 3 | 173 | 4685972 | ||
BC106003.1 | - | 3 | 137 | 6143473 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
