siRNA Details
Gene Symbol: RILPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319243.1 | NP_001306172.1 | 1 | 516 | ||
NM_178314.4 | NP_847884.2 | 1 | 516 | |||
XM_006719372.2 | XP_006719435.1 | 1 | 510 | |||
GenBank mRNA | AK096697.1 | - | 2 | 358 | ||
BC089444.1 | - | 1 | 318 | 30559974 | ||
BX281699.1 | - | 1 | 169 | |||
BX359997.2 | - | 1 | 516 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
