siRNA Details
Gene Symbol: RILPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319243.1 | NP_001306172.1 | 7 | 1341 | ||
GenBank mRNA | AK096697.1 | - | 8 | 1183 | ||
BC041375.1 | - | 1 | 379 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
