siRNA Details
Gene Symbol: LCE1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178349.1 | NP_848126.1 | 1 | 1065 | ||
GenBank mRNA | BC031811.1 | - | 1 | 1065 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LCE1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178349.1 | NP_848126.1 | 1 | 1065 | ||
GenBank mRNA | BC031811.1 | - | 1 | 1065 |