siRNA Details
Gene Symbol: LCE2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178430.3 | NP_848517.1 | 2 | 429 | ||
GenBank mRNA | BC130603.1 | - | 2 | 429 | 40146634 | |
BC130605.1 | - | 2 | 429 | 40146636 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LCE2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178430.3 | NP_848517.1 | 2 | 429 | ||
GenBank mRNA | BC130603.1 | - | 2 | 429 | 40146634 | |
BC130605.1 | - | 2 | 429 | 40146636 |