siRNA Details
Gene Symbol: FGF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC069692.1 | - | 3 | 577 | 7262620 | |
BC103978.1 | - | 3 | 471 | 40015567 | ||
BC103979.1 | - | 3 | 471 | 40015568 | ||
D14838.1 | - | 3 | 626 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
