siRNA Details
Gene Symbol: TMPRSS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182973.1 | NP_892018.1 | 9, 10 | 1463 | ||
XM_011527978.2 | XP_011526280.1 | 11, 12 | 1826 | |||
XM_011527980.1 | XP_011526282.1 | 3, 4 | 482 | |||
GenBank mRNA | AJ488946.1 | - | 9, 10 | 1463 | ||
AJ488947.1 | - | 9, 10 | 1463 | |||
AK131261.1 | - | 1, 2 | 52 |
