siRNA Details
Gene Symbol: FAM111B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142703.1 | NP_001136175.1 | 3 | 1433 | ||
NM_001142704.1 | NP_001136176.1 | 2 | 1388 | |||
NM_198947.3 | NP_945185.1 | 4 | 1600 | |||
GenBank mRNA | AK303520.1 | - | 3 | 1382 | ||
AK304258.1 | - | 4 | 1548 | |||
AY457926.1 | - | 4 | 1549 | |||
BC130513.1 | - | 3 | 1490 | 40146544 | ||
BC130539.1 | - | 3 | 1490 | 40146570 |
