siRNA Details
Gene Symbol: C12orf42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099336.2 | NP_001092806.1 | 5 | 753 | ||
NM_001278419.1 | NP_001265348.1 | 6 | 848 | |||
NM_001278420.1 | NP_001265349.1 | 6 | 713 | |||
NM_198521.3 | NP_940923.2 | 5 | 618 | |||
XM_006719377.3 | XP_006719440.1 | 4 | 556 | |||
XM_011538291.1 | XP_011536593.1 | 5 | 613 | |||
XM_011538292.2 | XP_011536594.1 | 5 | 582 | |||
XM_011538293.2 | XP_011536595.1 | 5 | 589 | |||
XM_011538294.2 | XP_011536596.1 | 5 | 563 | |||
XM_011538295.1 | XP_011536597.1 | 5 | 645 | |||
XM_011538297.2 | XP_011536599.1 | 6 | 1629 | |||
XM_011538300.1 | XP_011536602.1 | 6 | 798 | |||
XM_011538305.1 | XP_011536607.1 | 5 | 815 | |||
XM_017019276.1 | XP_016874765.1 | 5 | 707 | |||
XM_017019277.1 | XP_016874766.1 | 8 | 1082 | |||
XM_017019278.1 | XP_016874767.1 | 7 | 861 | |||
XM_017019279.1 | XP_016874768.1 | 8 | 1387 | |||
XM_017019280.1 | XP_016874769.1 | 8 | 1087 | |||
GenBank mRNA | AK058052.1 | - | 5 | 576 | ||
BC044617.1 | - | 6 | 809 | 5267756 | ||
BC117392.1 | - | 5 | 576 | 40125943 | ||
BC117394.1 | - | 5 | 576 | 40125945 | ||
BI830750.1 | - | 5 | 696 |
