siRNA Details
Gene Symbol: DRAXIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198545.3 | NP_940947.3 | 4 | 847 | ||
XM_011541405.2 | XP_011539707.1 | 5 | 1021 | |||
XM_017001214.1 | XP_016856703.1 | 5 | 1157 | |||
GenBank mRNA | AB301919.1 | - | 3 | 707 | ||
AK075558.1 | - | 4 | 842 | |||
AY358750.1 | - | 4 | 808 | |||
BC021286.2 | - | 2 | 197 | |||
BC111064.1 | - | 4 | 849 | 5769984 |
