siRNA Details
Gene Symbol: LOC105379272 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011544006.2 | XP_011542308.1 | 3 | 565 |
Gene Symbol: ANKRD20A12P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046228.1 | 4 | 1221 | |||
GenBank mRNA | AK090412.1 | - | 4 | 2222 |
Gene Symbol: LOC101059949 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017030105.1 | XP_016885594.1 | 2 | 921 |
Gene Symbol: LOC101927345 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011544009.2 | XP_011542311.1 | 2 | 444 |
