siRNA Details
Gene Symbol: MAST4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164664.1 | NP_001158136.1 | 1, 2 | 673 | ||
NM_001290228.1 | NP_001277157.1 | 1, 2 | 673 | |||
NM_198828.2 | NP_942123.1 | 1, 2 | 673 | |||
XM_011543382.2 | XP_011541684.1 | 1, 2 | 654 | |||
XM_017009447.1 | XP_016864936.1 | 1, 2 | 654 | |||
XM_017009448.1 | XP_016864937.1 | 1, 2 | 654 | |||
GenBank mRNA | AY439222.1 | - | 1, 2 | 365 | ||
BC033215.1 | - | 1, 2 | 641 | 5015614 | ||
CB044105.1 | - | 1, 2 | 573 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
