siRNA Details
Gene Symbol: NAT16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198571.2 | NP_940973.2 | 4 | 2082 | ||
XM_005250325.1 | XP_005250382.1 | 5 | 2061 | |||
XM_011516168.1 | XP_011514470.1 | 4 | 2043 | |||
GenBank mRNA | AK092119.1 | - | 3 | 1310 | ||
AK096556.1 | - | 4 | 2082 | |||
AL831911.1 | - | 2 | 1352 |
