siRNA Details
Gene Symbol: KCP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199349.2 | NP_955381.2 | 6 | 674 | ||
XM_017012184.1 | XP_016867673.1 | 6 | 674 | |||
XM_017012185.1 | XP_016867674.1 | 6 | 674 | |||
XM_017012186.1 | XP_016867675.1 | 6 | 674 | |||
XM_017012187.1 | XP_016867676.1 | 6 | 674 | |||
XM_017012188.1 | XP_016867677.1 | 6 | 674 | |||
XM_017012189.1 | XP_016867678.1 | 6 | 674 | |||
XM_017012190.1 | XP_016867679.1 | 6 | 674 | |||
XM_017012193.1 | XP_016867682.1 | 6 | 674 | |||
XM_017012194.1 | XP_016867683.1 | 6 | 674 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
