siRNA Details
Gene Symbol: SPATC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134374.1 | NP_001127846.1 | 3 | 1330 | ||
NM_198572.2 | NP_940974.2 | 3 | 1330 | |||
XM_011517021.1 | XP_011515323.1 | 3 | 1330 | |||
XM_011517022.2 | XP_011515324.1 | 3 | 1616 | |||
XM_011517023.2 | XP_011515325.1 | 3 | 2634 | |||
XM_011517024.2 | XP_011515326.1 | 4 | 1855 | |||
XM_011517025.2 | XP_011515327.1 | 3 | 2290 | |||
XM_017013399.1 | XP_016868888.1 | 4 | 1149 | |||
GenBank mRNA | AB092352.1 | - | 3 | 1318 | ||
AK301710.1 | - | 3 | 1318 | |||
BC050390.1 | - | 3 | 1320 | 5742710 | ||
BC053547.1 | - | 3 | 1305 | 5743874 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
