siRNA Details
Gene Symbol: WASH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182905.4 | NP_878908.4 | 7 | 989 | ||
XM_011517658.2 | XP_011515960.1 | 7 | 1106 | |||
XM_011517659.2 | XP_011515961.1 | 7 | 888 | |||
XM_011517660.2 | XP_011515962.1 | 7 | 885 | |||
XM_011517662.2 | XP_011515964.1 | 7 | 2392 | |||
XM_011517663.2 | XP_011515965.1 | 6 | 2241 | |||
XM_011517664.2 | XP_011515966.1 | 6 | 2238 | |||
XM_011517665.2 | XP_011515967.1 | 6 | 787 | |||
XM_011517666.2 | XP_011515968.1 | 6 | 786 | |||
XM_011517671.2 | XP_011515973.2 | 6 | 970 | |||
XM_017014169.1 | XP_016869658.1 | 7 | 1005 | |||
XM_017014170.1 | XP_016869659.1 | 7 | 1068 | |||
XM_017014171.1 | XP_016869660.1 | 5 | 680 | |||
XM_017014172.1 | XP_016869661.1 | 5 | 2258 | |||
XM_017014173.1 | XP_016869662.1 | 6 | 970 | |||
GenBank mRNA | AK024481.1 | - | 1 | 1843 |
Gene Symbol: LOC100288778 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130745.1 | 7 | 989 | |||
GenBank mRNA | BC063682.1 | - | 3 | 335 |
Gene Symbol: WASH2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024077.2 | 7 | 1114 | |||
GenBank mRNA | AK292759.1 | - | 6 | 717 | ||
AL137714.1 | - | 6 | 1240 | |||
AY341936.1 | - | 2 | 243 | |||
AY341937.1 | - | 2 | 243 | |||
BC073913.1 | - | 2 | 510 | |||
BC082240.1 | - | 4 | 1809 | |||
BC093083.1 | - | 2 | 523 | |||
BX537637.1 | - | 7 | 1115 |
Gene Symbol: WASH3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003659.2 | 7 | 1663 | |||
GenBank mRNA | AK056232.1 | - | 7 | 1508 | ||
AK057951.1 | - | 9 | 1113 | |||
AK095183.1 | - | 6 | 1109 | |||
AK290087.1 | - | 5 | 729 | |||
AK291582.1 | - | 6 | 1124 | |||
BC063555.1 | - | 6 | 1302 | |||
BC110991.1 | - | 2 | 276 | |||
BQ935819.1 | - | 2 | 279 | |||
DA065132.1 | - | 1 | 506 |
Gene Symbol: WASH7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024540.1 | 7 | 888 | |||
GenBank mRNA | AK092583.1 | - | 6 | 763 | ||
AK296182.1 | - | 8 | 1024 | |||
AK302089.1 | - | 7 | 942 | |||
AK308566.1 | - | 5 | 625 | |||
AK310121.1 | - | 7 | 762 | |||
AK310139.1 | - | 8 | 916 | |||
AK316487.1 | - | 6 | 1077 | |||
BC063470.1 | - | 4 | 801 | |||
BC094698.1 | - | 4 | 1663 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
