siRNA Details
Gene Symbol: WASH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182905.4 | NP_878908.4 | 9 | 1293 | ||
XM_011517658.2 | XP_011515960.1 | 9 | 1410 | |||
XM_011517659.2 | XP_011515961.1 | 9 | 1192 | |||
XM_011517660.2 | XP_011515962.1 | 9 | 1189 | |||
XM_011517662.2 | XP_011515964.1 | 9 | 2696 | |||
XM_011517663.2 | XP_011515965.1 | 8 | 2545 | |||
XM_011517664.2 | XP_011515966.1 | 8 | 2542 | |||
XM_011517665.2 | XP_011515967.1 | 8 | 1091 | |||
XM_011517666.2 | XP_011515968.1 | 8 | 1090 | |||
XM_017014169.1 | XP_016869658.1 | 9 | 1309 | |||
XM_017014170.1 | XP_016869659.1 | 9 | 1372 | |||
XM_017014171.1 | XP_016869660.1 | 7 | 984 | |||
XM_017014172.1 | XP_016869661.1 | 7 | 2562 | |||
GenBank mRNA | AK024481.1 | - | 2 | 2802 | ||
BC094860.1 | - | 1 | 334 | |||
CN404407.1 | - | 2 | 190 |
Gene Symbol: LOC100288778 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130745.1 | 9 | 1293 | |||
GenBank mRNA | AW881719.1 | - | 1 | 470 | ||
BC063682.1 | - | 5 | 639 |
Gene Symbol: WASH2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024077.2 | 9 | 1414 | |||
GenBank mRNA | AK292759.1 | - | 8 | 1021 | ||
AL137714.1 | - | 7 | 2196 | |||
AY341936.1 | - | 4 | 547 | |||
AY341937.1 | - | 4 | 547 | |||
BC073913.1 | - | 4 | 810 | |||
BC082240.1 | - | 6 | 2109 | |||
BC093083.1 | - | 4 | 823 | |||
BX537637.1 | - | 9 | 1415 |
Gene Symbol: WASH3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003659.2 | 9 | 1967 | |||
GenBank mRNA | AK056232.1 | - | 9 | 1812 | ||
AK057951.1 | - | 11 | 1417 | |||
AK095183.1 | - | 8 | 1413 | |||
AK290087.1 | - | 7 | 1033 | |||
AK291582.1 | - | 8 | 1428 | |||
BC063555.1 | - | 8 | 1602 | |||
BC110991.1 | - | 4 | 576 |
Gene Symbol: WASH6P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC029274.1 | - | 2 | 202 |
Gene Symbol: WASH7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024540.1 | 9 | 1187 | |||
GenBank mRNA | AK092583.1 | - | 7 | 1722 | ||
AK308566.1 | - | 7 | 925 | |||
BC063470.1 | - | 6 | 1101 | |||
BC094698.1 | - | 6 | 1963 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
