siRNA Details
Gene Symbol: WASH7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024540.1 | 7 | 778 | |||
GenBank mRNA | AK092583.1 | - | 6 | 653 | ||
AK294377.1 | - | 5 | 911 | |||
AK302089.1 | - | 7 | 832 | |||
AK308566.1 | - | 5 | 515 | |||
AK310121.1 | - | 7 | 652 | |||
AK310139.1 | - | 8 | 806 | |||
AK316487.1 | - | 6 | 967 | |||
BC063470.1 | - | 4 | 691 | |||
BC094698.1 | - | 4 | 1553 |
Gene Symbol: LOC100288778 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130745.1 | 7 | 879 | |||
GenBank mRNA | BC063682.1 | - | 3 | 225 |
Gene Symbol: WASH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK024481.1 | - | 1 | 1733 |
Gene Symbol: WASH2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024077.2 | 7 | 1004 | |||
GenBank mRNA | AK292759.1 | - | 6 | 607 | ||
AK308574.1 | - | 6 | 693 | |||
AL137714.1 | - | 6 | 1130 | |||
AY341936.1 | - | 2 | 133 | |||
AY341937.1 | - | 2 | 133 | |||
BC073913.1 | - | 2 | 400 | |||
BC082240.1 | - | 4 | 1699 | |||
BC093083.1 | - | 2 | 413 | |||
BX377099.2 | - | 6 | 631 | |||
BX537637.1 | - | 7 | 1005 |
Gene Symbol: WASH3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003659.2 | 7 | 1553 | |||
GenBank mRNA | AK056232.1 | - | 7 | 1398 | ||
AK057951.1 | - | 9 | 1003 | |||
AK095183.1 | - | 6 | 999 | |||
AK290087.1 | - | 5 | 619 | |||
AK291582.1 | - | 6 | 1014 | |||
BC063555.1 | - | 6 | 1192 | |||
BC110991.1 | - | 2 | 166 | |||
BQ935819.1 | - | 2 | 169 | |||
DA065132.1 | - | 1 | 396 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
