siRNA Details
Gene Symbol: SH2D4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145719.1 | NP_001139191.1 | 3 | 415 | ||
NM_207372.2 | NP_997255.2 | 3 | 914 | |||
XM_017016236.1 | XP_016871725.1 | 3 | 3329 | |||
GenBank mRNA | AK123885.1 | - | 3 | 914 | ||
AK123978.1 | - | 3 | 415 |
