siRNA Details
Gene Symbol: SPATA41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028139.1 | 1 | 789 | |||
NR_028140.1 | 1, 2 | 304 | ||||
GenBank mRNA | AK124283.1 | - | 1 | 784 | ||
AY605659.1 | - | 1 | 43 | |||
BC028144.1 | - | 1, 2 | 300 | 5214282 | ||
BC132704.1 | - | 1, 2 | 247 | |||
BC132706.1 | - | 1, 2 | 247 |
