siRNA Details
Gene Symbol: SHISA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173461.1 | NP_001166932.1 | 4 | 1352 | ||
NM_001173462.1 | NP_001166933.1 | 5 | 1448 | |||
NM_207386.3 | NP_997269.2 | 6 | 1505 | |||
XM_017024618.1 | XP_016880107.1 | 5 | 1409 | |||
GenBank mRNA | AK127379.1 | - | 4 | 1046 | ||
AK128003.1 | - | 5 | 848 | |||
DQ070854.1 | - | 4 | 824 |
