siRNA Details
Gene Symbol: SHISA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173461.1 | NP_001166932.1 | 4 | 972 | ||
NM_001173462.1 | NP_001166933.1 | 5 | 1068 | |||
NM_207386.3 | NP_997269.2 | 6 | 1125 | |||
XM_017024618.1 | XP_016880107.1 | 5 | 1029 | |||
GenBank mRNA | AK127379.1 | - | 4 | 666 | ||
AK128003.1 | - | 5 | 468 |
