siRNA Details
Gene Symbol: ZNF808 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005258909.3 | XP_005258966.1 | 4 | 3338 | ||
GenBank mRNA | BC017109.1 | - | 2 | 592 | ||
BC033230.1 | - | 2 | 592 | |||
BC130351.1 | - | 1 | 466 | |||
BC130353.1 | - | 1 | 466 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
