siRNA Details
Gene Symbol: ZNF808 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039886.3 | NP_001034975.2 | 5 | 3182 | ||
NM_001321424.1 | NP_001308353.1 | 5 | 3327 | |||
NM_001321425.1 | NP_001308354.1 | 4 | 3080 | |||
XM_005258909.3 | XP_005258966.1 | 4 | 3043 | |||
GenBank mRNA | BC017109.1 | - | 2 | 297 | ||
BC033230.1 | - | 2 | 297 | |||
BC130351.1 | - | 1 | 171 | |||
BC130353.1 | - | 1 | 171 | |||
CR749856.1 | - | 3 | 2958 | |||
FJ795029.1 | - | 1 | 120 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
