siRNA Details
Gene Symbol: ZNF888 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310127.1 | NP_001297056.1 | 3 | 911 | ||
XM_005259451.4 | XP_005259508.1 | 5 | 1143 | |||
XM_017026798.1 | XP_016882287.1 | 5 | 1304 | |||
XM_017026799.1 | XP_016882288.1 | 5 | 1189 | |||
XM_017026800.1 | XP_016882289.1 | 3 | 920 |
