siRNA Details
Gene Symbol: C1orf146 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012425.1 | NP_001012425.1 | 2, 3 | 133 | ||
XM_011541447.2 | XP_011539749.1 | 3, 4 | 328 | |||
GenBank mRNA | BC130499.1 | - | 2, 3 | 127 | 40146530 | |
BU567193.1 | - | 2, 3 | 212 |
