siRNA Details
Gene Symbol: C1orf53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024594.2 | NP_001019765.1 | 2 | 379 | ||
GenBank mRNA | BC038214.1 | - | 2 | 278 | ||
BC130608.1 | - | 2 | 379 | 40146639 | ||
BC130634.1 | - | 2 | 379 | 40146665 |
