siRNA Details
Gene Symbol: TSPYL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003937.2 | NP_001003937.2 | 1 | 815 | ||
GenBank mRNA | AK097417.1 | - | 1 | 769 | ||
BC068576.1 | - | 1 | 750 | 4374337 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TSPYL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003937.2 | NP_001003937.2 | 1 | 815 | ||
GenBank mRNA | AK097417.1 | - | 1 | 769 | ||
BC068576.1 | - | 1 | 750 | 4374337 |