siRNA Details
Gene Symbol: TMEM110-MUSTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL833659.1 | - | 4 | 5108 |
Gene Symbol: MUSTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_205853.3 | NP_995325.3 | 3 | 569 | ||
GenBank mRNA | AJ276556.1 | - | 3 | 414 | ||
BC130550.1 | - | 3 | 340 | 40146581 | ||
BC130552.1 | - | 3 | 338 | 40146583 | ||
BX089772.1 | - | 3 | 554 | |||
CB043913.1 | - | 3 | 579 | |||
CR990075.1 | - | 3 | 460 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
