siRNA Details
Gene Symbol: C4orf50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011513603.2 | XP_011511905.1 | 8 | 3159 | ||
XM_011513604.2 | XP_011511906.1 | 9 | 3425 | |||
XM_017008891.1 | XP_016864380.1 | 10 | 3958 | |||
XM_017008892.1 | XP_016864381.1 | 10 | 3958 | |||
XM_017008893.1 | XP_016864382.1 | 9 | 3791 | |||
GenBank mRNA | AK128339.1 | - | 6 | 563 | ||
BC140710.1 | - | 6 | 539 | 9021578 |
