siRNA Details
Gene Symbol: ARHGEF37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001669.2 | NP_001001669.2 | 4 | 458 | ||
XM_005268447.4 | XP_005268504.1 | 4 | 429 | |||
XM_005268448.1 | XP_005268505.1 | 3 | 332 | |||
XM_006714784.3 | XP_006714847.1 | 5 | 797 | |||
XM_011537642.2 | XP_011535944.1 | 4 | 480 | |||
XM_017009460.1 | XP_016864949.1 | 4 | 682 | |||
XM_017009461.1 | XP_016864950.1 | 3 | 332 | |||
GenBank mRNA | AB199796.1 | - | 3 | 405 | ||
AK300355.1 | - | 3 | 319 | |||
AK300559.1 | - | 4 | 557 | |||
AK316002.1 | - | 4 | 528 | |||
BC041325.1 | - | 4 | 458 |
