siRNA Details
Gene Symbol: SPDYE8P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003664.2 | 8 | 2225 | |||
GenBank mRNA | BC045636.1 | - | 8 | 2228 | ||
BC068610.1 | - | 8 | 2228 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SPDYE8P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003664.2 | 8 | 2225 | |||
GenBank mRNA | BC045636.1 | - | 8 | 2228 | ||
BC068610.1 | - | 8 | 2228 |