siRNA Details
Gene Symbol: FAM90A25P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073395.1 | 4 | 1701 | |||
GenBank mRNA | AL832996.1 | - | 4 | 1700 |
Gene Symbol: FAM90A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319982.1 | NP_001306911.1 | 6 | 2161 | ||
NM_018088.3 | NP_060558.3 | 7 | 2318 | |||
XM_006719097.3 | XP_006719160.1 | 3 | 2222 | |||
XM_011520719.1 | XP_011519021.1 | 7 | 2309 | |||
XM_017019547.1 | XP_016875036.1 | 7 | 2306 | |||
GenBank mRNA | AK001270.1 | - | 6 | 2147 | ||
BC042608.1 | - | 7 | 2297 | 5167203 |
Gene Symbol: FAM90A7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046343.1 | 3 | 1646 | |||
GenBank mRNA | AK301810.1 | - | 4 | 1647 |
