siRNA Details
Gene Symbol: MROH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207414.2 | NP_997297.2 | 8 | 970 | ||
NR_102363.2 | 7 | 788 | ||||
NR_102364.2 | 8 | 929 | ||||
GenBank mRNA | AK125848.1 | - | 8 | 970 | ||
AK131461.1 | - | 8 | 929 | |||
BC144560.1 | - | 7 | 764 |
