siRNA Details
Gene Symbol: FAM205A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001141917.1 | NP_001135389.1 | 4 | 3741 | ||
GenBank mRNA | AK125949.1 | - | 2 | 2719 | ||
AK302320.1 | - | 3 | 2094 |
Gene Symbol: FAM205BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024481.1 | 4 | 3724 | |||
GenBank mRNA | AK128180.1 | - | 4 | 3724 | ||
AL080137.1 | - | 1 | 975 | |||
BX648118.1 | - | 4 | 3669 |
