siRNA Details
Gene Symbol: CFAP77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282957.1 | NP_001269886.1 | 1 | 245 | ||
NM_207417.2 | NP_997300.1 | 1 | 245 | |||
XM_011518670.2 | XP_011516972.1 | 1 | 398 | |||
XM_011518671.1 | XP_011516973.1 | 1 | 398 | |||
XM_017014711.1 | XP_016870200.1 | 1 | 398 | |||
GenBank mRNA | AK128819.1 | - | 1 | 217 | ||
BC118593.1 | - | 1 | 213 | 40108008 | ||
BC124555.1 | - | 1 | 213 | 40108012 | ||
HY017665.1 | - | 1 | 246 |
