siRNA Details
Gene Symbol: OR51M1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004756.2 | NP_001004756.2 | 1 | 188 | ||
GenBank mRNA | BC136912.1 | - | 1 | 188 | 9020903 | |
BC136913.1 | - | 1 | 188 | 9020904 |
