siRNA Details
Gene Symbol: OR51I1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005288.2 | NP_001005288.1 | 1 | 675 | ||
GenBank mRNA | BC151148.1 | - | 1 | 675 | 9021023 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: OR51I1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005288.2 | NP_001005288.1 | 1 | 675 | ||
GenBank mRNA | BC151148.1 | - | 1 | 675 | 9021023 |