siRNA Details
Gene Symbol: OR9G1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005213.1 | NP_001005213.1 | 1 | 212 |
Gene Symbol: OR9G9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013358.2 | NP_001013376.2 | 1 | 212 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_146568, BC111581.
Set Membership: |
![]() |
Gene Ontology Categories:
