siRNA Details
Gene Symbol: OR4M2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004719.2 | NP_001004719.2 | 1 | 949 | ||
GenBank mRNA | BC136985.1 | - | 1 | 949 | 9020977 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: OR4M2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004719.2 | NP_001004719.2 | 1 | 949 | ||
GenBank mRNA | BC136985.1 | - | 1 | 949 | 9020977 |