siRNA Details
Gene Symbol: NAGS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153006.2 | NP_694551.1 | 7 | 1781 | ||
XM_011524438.1 | XP_011522740.1 | 6 | 1716 | |||
XM_011524439.2 | XP_011522741.1 | 7 | 1671 | |||
GenBank mRNA | AK074407.1 | - | 7 | 1711 | ||
AK091938.1 | - | 4 | 1669 |
