siRNA Details
Gene Symbol: ZNF793 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013659.2 | NP_001013681.2 | 8 | 1053 | ||
XM_005258927.3 | XP_005258984.1 | 8 | 1165 | |||
XM_006723213.3 | XP_006723276.1 | 7 | 1075 | |||
GenBank mRNA | AK131417.1 | - | 8 | 1053 | ||
AK303740.1 | - | 6 | 846 | |||
BX537524.1 | - | 8 | 1529 |
