siRNA Details
Gene Symbol: OR10K1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004473.1 | NP_001004473.1 | 1 | 484 | ||
GenBank mRNA | BC137323.1 | - | 1 | 564 | 9021320 | |
BC137324.1 | - | 1 | 564 | 9021321 |
